1-Brom-5-fluor-2-nitrobenzol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Brom-4-fluornitrobenzol, 1-Brom-3-fluor-6-nitrobenzol
H61466.06
H61466.14
H61466.06EA
53
EUR
H61466.06
H61466.14
1-Brom-5-fluor-2-nitrobenzol ≥98%
1-Brom-5-fluor-2-nitrobenzol
Formel:
C₆H₃BrFNO₂ Molecular Weight: 220 g/mol Schmelzpunkt: 41…45 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00792441 CAS-Nummer: 700-36-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...