1-Brom-2-fluor-4-nitrobenzol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

H61494.14 H61494.22
H61494.14EA 114 EUR
H61494.14 H61494.22
1-Brom-2-fluor-4-nitrobenzol ≥98%
1-Bromo-2-fluoro-4-nitrobenzene
Formel: C₆H₃BrFNO₂
Schmelzpunkt: 88…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05865088
CAS-Nummer: 185331-69-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR