7-Fluoroxindol 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 7-Fluoroindolin-2-one

H61511.MD H61511.03 H61511.06
H61511.MDEA 45.9 EUR
H61511.MD H61511.03 H61511.06
7-Fluoroxindol 95%
7-Fluoroxindol
Formel: C₈H₆FNO
Molecular Weight: 151.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02179608
CAS-Nummer: 71294-03-6
EINECS: 676-237-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR