3-Brom-5-methylanilin 98%

Lieferant: Thermo Fisher Scientific
Achtung

H61519.03 H61519.06
H61519.03EA 62.5 EUR
H61519.03 H61519.06
3-Brom-5-methylanilin 98%
3-Bromo-5-methylaniline

Purity: 98% CAS No.: 74586-53-1 Molecular Formula: C7H8BrN Formula Weight: 186.05 Physical Form: Powder Appearance: Brown UN No.: UN2811 MDL No.: MFCD11845553

Formel: C7H8BrN
Molecular Weight: 186.05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11845553
CAS-Nummer: 74586-53-1
EINECS: 277-932-1
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR