4-Fluorbenzamidin Hydrochlorid 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H61570.MD
H61570.MDEA
0
EUR
H61570.MD
4-Fluorbenzamidin Hydrochlorid 95%
4-Fluorbenzamidin Hydrochlorid
Formel:
C₇H₈ClFN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04114421 CAS-Nummer: 456-14-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...