2-Cyan-6-fluoranilin 95%

Lieferant: Thermo Fisher Scientific
Achtung

H61580.MD H61580.03
H61580.MDEA 44.5 EUR
H61580.MD H61580.03
2-Cyan-6-fluoranilin 95%
2-Cyan-6-fluoranilin
Formel: C₇H₅FN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07368774
CAS-Nummer: 115661-37-5
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR