2-Brom-5-fluorbenzylalkohol ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (2-Bromo-5-fluorophenyl)methanol

H61596.06
H61596.06EA 176 EUR
H61596.06
2-Brom-5-fluorbenzylalkohol ≥97%
2-Brom-5-fluorbenzylalkohol
Formel: C₇H₆BrFO
Molecular Weight: 205,03 g/mol
Schmelzpunkt: 91…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00142876
CAS-Nummer: 202865-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR