2-Biphenylyl(diphenyl)phosphin 97%

Lieferant: Thermo Scientific
Achtung

H61640.06
H61640.06EA 88.2 EUR
H61640.06
2-Biphenylyl(diphenyl)phosphin 97%
2-(Diphenylphosphino)biphenyl
Formel: C₂₄H₁₉P
Molecular Weight: 338,38 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11559063
CAS-Nummer: 13885-09-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR