5-Brom-2-fluoranilin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H61678.14
H61678.22
H61678.14EA
74.9
EUR
H61678.14
H61678.22
5-Brom-2-fluoranilin ≥98%
Purity: 98%
CAS No.: 2924-09-6
Molecular Formula: C6H5BrFN
Formula Weight: 190.01
Physical Form: Solid
Melting Point: 27°
Boiling Point: 105°/12mm
Beilstein Reference: 1241528
UN No.: 2811
MDL No.: MFCD03094181
Formel:
C₆H₅BrFN Siedepunkt: 105 °C (12 mmHg) Schmelzpunkt: 27 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094181 CAS-Nummer: 2924-09-6 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...