4-Fluor-4'-methoxybenzophenon ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H61679.MD
H61679.MDEA
0
EUR
H61679.MD
4-Fluor-4'-methoxybenzophenon ≥98%
4-Fluor-4'-methoxybenzophenon
Formel:
C₁₄H₁₁FO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00055469 CAS-Nummer: 345-89-1 EINECS: 206-464-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...