Ethyl-6-chlorpicolinat ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl 6-chloropicolinate, Ethyl 6-chloropyridine-2-carboxylate

H61701.06
H61701.06EA 121 EUR
H61701.06
Ethyl-6-chlorpicolinat ≥97%
Ethyl-6-chlorpicolinat
Formel: C₈H₈ClNO₂
Molecular Weight: 185,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878438
CAS-Nummer: 21190-89-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR