3-Brom-4-fluorphenol 97%
Lieferant: Thermo Scientific
|
Achtung
|
H61702.06
H61702.06EA
88
EUR
H61702.06
3-Brom-4-fluorphenol 97%
3-Brom-4-fluorphenol
Purity: 98%
CAS No.: 27407-11-0
Molecular Formula: C6H4BrFO
Formula Weight: 191.00
Physical Form: Crystalline
Appearance: Yellow-green
UN No.: 2811
MDL No.: MFCD03425884
Formel:
C₆H₄BrFO Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425884 CAS-Nummer: 27407-11-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...