Methyl-2-amino-4-fluorbenzoat 97%

Lieferant: Thermo Scientific

Synonyme: Methyl 2-amino-4-fluorobenzoate

H61749.14
H61749.14EA 193 EUR
H61749.14
Methyl-2-amino-4-fluorbenzoat 97%
Methyl-2-amino-4-fluorbenzoat
Formel: C₈H₈FNO₂
Molecular Weight: 169,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11054082
CAS-Nummer: 2475-81-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR