2,6-Dichlor-4-methylpyridin 95%

Lieferant: Thermo Scientific
Gefahr

H61767.03
H61767.03EA 85.1 EUR
H61767.03
2,6-Dichlor-4-methylpyridin 95%
2,6-Dichloro-4-methylpyridine
Formel: C₆H₅Cl₂N
Molecular Weight: 162,02 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD09264302
CAS-Nummer: 39621-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR