5-Fluor-2-nitroanilin 97%

Lieferant: Thermo Fisher Scientific
Gefahr

H61806.06 H61806.14 H61806.22
H61806.06EA 36.1 EUR
H61806.06 H61806.14 H61806.22
5-Fluor-2-nitroanilin 97%
5-Fluor-2-nitroanilin
Formel: C₆H₅FN₂O₂
Molecular Weight: 156.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00034065
CAS-Nummer: 2369-11-1
EINECS: 219-128-5
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR