2-Amino-5-brom-2'-fluorbenzophenon 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: (2-Amino-5-bromophenyl)(2-fluorophenyl)methanone, 2-Amino-2'-fluoro-5-bromobenzophenone

H61882.06
H61882.06EA 75 EUR
H61882.06
2-Amino-5-brom-2'-fluorbenzophenon 95%
2-Amino-5-brom-2'-fluorbenzophenon
Formel: C₁₃H₉BrFNO
Molecular Weight: 294,12 g/mol
Schmelzpunkt: 100…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00038380
CAS-Nummer: 1479-58-9
EINECS: 216-037-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR