5'-Brom-2'-hydroxyacetophenon ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-(5-Brom-2-hydroxyphenyl)ethan-1-on, 2-Acetyl-4-bromphenol
H61979.09
H61979.18
H61979.09EA
45.8
EUR
H61979.09
H61979.18
5'-Brom-2'-hydroxyacetophenon ≥98%
5'-Brom-2'-hydroxyacetophenon
Formel:
C₈H₇BrO₂ Schmelzpunkt: 56…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191850 CAS-Nummer: 1450-75-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...