1-Chlor-2-fluor-3-nitrobenzol ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3-Chloro-2-fluoronitrobenzene
H61989.14
H61989.14EA
79.8
EUR
H61989.14
1-Chlor-2-fluor-3-nitrobenzol ≥97%
1-Chloro-2-fluoro-3-nitrobenzene
Formel:
C₆H₃ClFNO₂ Molecular Weight: 175,55 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00069417 CAS-Nummer: 2106-49-2 EINECS: 218-285-7 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...