Boc-D-aspartic acid β-benzyl ester 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-Boc-D-aspartic acid 4-benzyl ester, Boc-D-Asp(OBzl)-OH
H62000.14
H62000.14EA
227
EUR
H62000.14
Boc-D-aspartic acid β-benzyl ester 98%
Boc-D-aspartic acid beta-benzyl ester
Purity: 98%
CAS No.: 51186-58-4
Molecular Formula: C16H21NO6
Formula Weight: 323.34
Physical Form: Solid
Beilstein Reference: 2305471
MDL No.: MFCD00038255
Formel:
C₁₆H₂₁NO₆ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038255 CAS-Nummer: 51186-58-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...