(2-Brom-5-fluorphenyl)essigsäure 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Bromo-5-fluorophenylacetic acid

H62005.06
H62005.06EA 391 EUR
H62005.06
(2-Brom-5-fluorphenyl)essigsäure 96%
(2-Brom-5-fluorphenyl)essigsäure

Purity: 96% CAS No.: 739336-26-6 Molecular Formula: C8H6BrFO2 Formula Weight: 233.03 Physical Form: Solid Melting Point: 113-114° MDL No.: MFCD03094440

Formel: C₈H₆BrFO₂
Molecular Weight: 233,03 g/mol
Schmelzpunkt: 113…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094440
CAS-Nummer: 739336-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR