6-Bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-Bromo-7-fluoro-2H-benzo[b][1,4]oxazin-3(4H)-one

H62026.03
H62026.03EA 283 EUR
H62026.03
6-Bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one 96%
6-Bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one

Purity: 96% CAS No.: 355423-58-4 Molecular Formula: C8H5BrFNO2 Formula Weight: 246.03 Physical Form: Solid MDL No.: MFCD20441932

Formel: C₈H₅BrFNO₂
Molecular Weight: 246,035 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20441932
CAS-Nummer: 355423-58-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR