4'-Brom-3'-fluoracetophenon 96%

Lieferant: Thermo Scientific

Synonyme: 4-Bromo-3-fluoro--acetophenone

H62040.06
H62040.06EA 428 EUR
H62040.06
4'-Brom-3'-fluoracetophenon 96%
4'-Brom-3'-fluoracetophenon

Purity: 96% CAS No.: 304445-49-6 Molecular Formula: C8H6BrFO Formula Weight: 217.04 Physical Form: Solid

Formel: C₈H₆BrFO
Molecular Weight: 217,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00466241
CAS-Nummer: 304445-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR