2,6-Difluoro-4-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-Yl)Phenol 96%
Lieferant: Thermo Fisher Scientific
Synonyme:
3,5-Difluoro-4-hydroxybenzeneboronic acid pinacol ester
H62042.MD
H62042.MDEA
106
EUR
H62042.MD
2,6-Difluoro-4-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-Yl)Phenol 96%
2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Purity: 96%
CAS No.: 1029439-83-5
Molecular Formula: C12H15BF2O3
Formula Weight: 256.05
Physical Form: Solid
Appearance: Off-white
MDL No.: MFCD16996286
Formel:
C₁₂H₁₅BF₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD16996286 CAS-Nummer: 1029439-83-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...