4-Fluoro-2-nitrobenzeneboronic acidpinacol ester ≥96%

Lieferant: Thermo Scientific

Synonyme: 2-(4-Fluoro-2-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H62059.03
H62059.03EA 268 EUR
H62059.03
4-Fluoro-2-nitrobenzeneboronic acidpinacol ester ≥96%
4-Fluoro-2-nitrobenzeneboronic acidpinacol ester

Purity: 96% CAS No.: 1288978-82-4 Molecular Formula: C12H15BFNO4 Formula Weight: 267.06 Physical Form: Solid

Formel: C₁₂H₁₅BFNO₄
Molecular Weight: 267,063 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD16996295
CAS-Nummer: 1288978-82-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR