trans-1-(Boc-amino)-4-cyanocyclohexane 97%

Lieferant: Thermo Fisher Scientific
Achtung

H62060.03
H62060.03EA 219 EUR
H62060.03
trans-1-(Boc-amino)-4-cyanocyclohexane 97%
trans-1-(Boc-amino)-4-cyanocyclohexane

Purity: 97% CAS No.: 873537-32-7 Molecular Formula: C12H20N2O2 Formula Weight: 224.29 UN No.: UN3439 MDL No.: MFCD11707221

Formel: C₁₂H₂₀N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11707221
CAS-Nummer: 873537-32-7
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR