Boc-Lys(boc)-OSu 97%

Lieferant: Thermo Fisher Scientific

H62068.06
H62068.06EA 117 EUR
H62068.06
Boc-Lys(boc)-OSu 97%
Boc-Lys(boc)-OSu

Purity: 97% CAS No.: 30189-36-7 Molecular Formula: C20H33N3O8 Formula Weight: 443.49 Storage Temperature: -30°C to -10°C Physical Form: Powder Sensitivity: Moisture Sensitive Appearance: Off-white Melting Point: 184° Beilstein Reference: 1559007 Optical Rotation: -25°(c=1 in DMF) MDL No.: MFCD00057898

Formel: C₂₀H₃₃N₃O₈
Schmelzpunkt: 184 °C
MDL: MFCD00057898
CAS-Nummer: 30189-36-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR