4-Cyanomethyl-3-fluorobenzeneboronic acid pinacol ester 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Cyanomethyl-3-fluorophenylboronic acid pinacol ester, [2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile, Benzeneacetonitrile, 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-

H62103.MD
H62103.MDEA 118 EUR
H62103.MD
4-Cyanomethyl-3-fluorobenzeneboronic acid pinacol ester 96%
4-Cyanomethyl-3-fluorobenzeneboronic acid pinacol ester

Purity: 96% Molecular Formula: C14H17BFNO2 Formula Weight: 261.10 Physical Form: Solid

Formel: C₁₄H₁₇BFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18383797
CAS-Nummer: 1220219-61-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR