Boc-D-Asp-OBzl 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-Boc-D-aspartic acid 1-benzyl ester
H62210.03
H62210.06
H62210.03EA
88.4
EUR
H62210.03
H62210.06
Boc-D-Asp-OBzl 98%
Boc-D-Asp-OBzl
Purity: 98%
CAS No.: 92828-64-3
Molecular Formula: C16H21NO6
Formula Weight: 323.34
Physical Form: Powder
Appearance: White
MDL No.: MFCD00038264
Formel:
C₁₆H₂₁NO₆ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038264 CAS-Nummer: 92828-64-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...