3-Carboxy-5-fluorphenylboronsäurepinakolester 96%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 3-Carboxy-5-fluorobenzeneboronic acid pinacol ester
H62225.03
H62225.03EA
398
EUR
H62225.03
3-Carboxy-5-fluorphenylboronsäurepinakolester 96%
3-Carboxy-5-fluorphenylboronsäurepinakolester
Purity: 96%
CAS No.: 936728-22-2
Molecular Formula: C13H16BFO4
Formula Weight: 266.07
Physical Form: Solid
Formel:
C₁₃H₁₆BFO₄ Molecular Weight: 266,07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12756465 CAS-Nummer: 936728-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...