3-Fluoro-5-hydroxybenzeneboronic acid pinacol ester 96%
Lieferant: Thermo Scientific
Synonyme:
5-Fluoro-3-hydroxyphenylboronic acid pinacol ester
H62234.03
H62234.03EA
510
EUR
H62234.03
3-Fluoro-5-hydroxybenzeneboronic acid pinacol ester 96%
3-Fluoro-5-hydroxybenzeneboronic acid pinacol ester
Purity: 96%
Molecular Formula: C12H16BFO3
Formula Weight: 238.06
Physical Form: Solid
Formel:
C₁₂H₁₆BFO₃ Molecular Weight: 238,06 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16994409 CAS-Nummer: 1887126-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...