4-Acetyl-2-fluorobenzeneboronic acid pinacol ester 96%

Lieferant: Thermo Fisher Scientific

Synonyme: 4-Acetyl-2-fluorophenylboronic acid pinacol ester, 1-(3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethan-1-one

H62256.03
H62256.03EA 292 EUR
H62256.03
4-Acetyl-2-fluorobenzeneboronic acid pinacol ester 96%
4-Acetyl-2-fluorobenzeneboronic acid pinacol ester

Purity: 96% Molecular Formula: C14H18BFO3 Formula Weight: 264.10 Physical Form: Solid

Formel: C₁₄H₁₈BFO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18732910
CAS-Nummer: 1807685-51-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR