Fmoc-L-homoleucine 95%

Lieferant: Thermo Scientific

Synonyme: Fmoc-Homoleu-OH, N-Fmoc-L-homoleucine, Fmoc-HoLeu-OH

H62270.03
H62270.03EA 191 EUR
H62270.03
Fmoc-L-homoleucine 95%
Fmoc-L-homoleucine

Purity: 95% CAS No.: 180414-94-2 Molecular Formula: C22H25NO4 Formula Weight: 367.44 Physical Form: Solid MDL No.: MFCD00270206

Formel: C₂₂H₂₅NO₄
Molecular Weight: 367,445 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00270206
CAS-Nummer: 180414-94-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR