(S)-N-tert-Butoxycarbonylalanine methyl ester 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
Boc-Ala-OMe, N-Boc-L-alanine methyl ester
H62271.06
H62271.14
H62271.06EA
59.4
EUR
H62271.06
H62271.14
(S)-N-tert-Butoxycarbonylalanine methyl ester 95%
(S)-N-tert-Butoxycarbonylalanine methyl ester
Purity: 95%
CAS No.: 28875-17-4
Molecular Formula: C9H17NO4
Formula Weight: 203.24
Flash Point: 113°(235°F)
Physical Form: Powder
Density: 1.030
Appearance: White
Melting Point: 32-35°
Beilstein Reference: 1872545
Optical Rotation: -45°(c=1 in methanol)
MDL No.: MFCD00038513
Formel:
C₉H₁₇NO₄ Molecular Weight: 203.24 g/mol Schmelzpunkt: 32…35 °C Dichte: 1.030 Flammpunkt: 113 °C (235 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038513 CAS-Nummer: 28875-17-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...