3,4-Dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester

Lieferant: Thermo Fisher Scientific

Synonyme: 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-benzo[b][1,4]oxazine

H62290.03
H62290.03EA 593 EUR
H62290.03
3,4-Dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester
3,4-Dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester

CAS No.: 1155264-46-2 Molecular Formula: C14H20BNO3 Formula Weight: 261.12 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD18073255

Formel: C₁₄H₂₀BNO₃
MDL: MFCD18073255
CAS-Nummer: 1155264-46-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR