N-Boc-sarcosine methyl ester 97%

Lieferant: Thermo Scientific

Synonyme: Boc-Gly-Ome, N-Boc-N-methylglycine methyl ester, Boc-Sar-OMe, Methyl 2-((tert-butoxycarbonyl)(methyl)amino)acetate

H62304.06
H62304.06EA 308 EUR
H62304.06
N-Boc-sarcosine methyl ester 97%
N-Boc-sarcosine methyl ester

Purity: 97% CAS No.: 42492-57-9 Molecular Formula: C9H17NO4 Formula Weight: 203.24 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Yellow MDL No.: MFCD04973115

Formel: C₉H₁₇NO₄
Molecular Weight: 203,24 g/mol
MDL: MFCD04973115
CAS-Nummer: 42492-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR