D-Alanin-Methylester Hydrochlorid 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
D-Alanine Methyl Ester Hydrochloride, (R)-2-Aminopropionic acid Methyl Ester Hydrochloride, H-D-Ala-OMe.HCl, H-D-Ala-OMe·HCl, Methyl D-alaninate hydrochloride, Methyl (R)-2-Aminopropionate Hydrochloride, (R)-2-Aminopropionic Acid Methyl Ester Hydrochloride, (R)-Methyl 2-aminopropionate hydrochloride
Purity: 98%
CAS No.: 14316-06-4
Molecular Formula: C4H9NO2.HCl
Formula Weight: 139.58
Physical Form: Crystalline powder
Appearance: White to off-white
Melting Point: 108-110°
Beilstein Reference: 4235975
Optical Rotation: -8°(c=2 in methanol)
MDL No.: MFCD00066141
Formel:
C₄H₉NO₂ Schmelzpunkt: 108…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066141 CAS-Nummer: 14316-06-4 EINECS: 238-258-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...