5-Brom-4-fluor-2-methylanilin 96%

Lieferant: Thermo Scientific
Achtung

H62480.06
H62480.06EA 206 EUR
H62480.06
5-Brom-4-fluor-2-methylanilin 96%
5-Brom-4-fluor-2-methylanilin

Purity: 96% CAS No.: 627871-16-3 Molecular Formula: C7H7BrFN Formula Weight: 204.04 Physical Form: Solid Melting Point: 86-90° MDL No.: MFCD05865218

Formel: C₇H₇BrFN
Molecular Weight: 204,04 g/mol
Schmelzpunkt: 86…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05865218
CAS-Nummer: 627871-16-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR