Boc-D-leucinol 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-Boc-D-leucinol
H62498.03
H62498.03EA
71.2
EUR
H62498.03
Boc-D-leucinol 98%
Boc-D-leucinol
Purity: 98%
CAS No.: 106930-51-2
Molecular Formula: C11H23NO3
Formula Weight: 217.31
Flash Point: 110°(230°F)
Physical Form: Liquid
Density: 0.983
Refractive Index: 1.4490
Appearance: Colourless
Boiling Point: 213°
Optical Rotation: +26°(c=2 in methanol)
MDL No.: MFCD00235931
Formel:
C₁₁H₂₃NO₃ Siedepunkt: 213 °C Dichte: 0,983 Flammpunkt: 110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00235931 CAS-Nummer: 106930-51-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...