Boc-Arg(Mtr)-OH 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Nalpha-Boc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulphonyl)-L-arginine
H62515.06
H62515.06EA
88.5
EUR
H62515.06
Boc-Arg(Mtr)-OH 98%
Boc-Arg(Mtr)-OH
Purity: 98%
CAS No.: 102185-38-6
Molecular Formula: C21H34N4O7S
Formula Weight: 486.53
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-ehite
MDL No.: MFCD00043097
Formel:
C₂₁H₃₄N₄O₇S |
MDL:
MFCD00043097 CAS-Nummer: 102185-38-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...