N-Boc-3-cyano-L-phenylalanine 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: Boc-Phe(3-CN)-OH

H62517.03
H62517.03EA 236 EUR
H62517.03
N-Boc-3-cyano-L-phenylalanine 95%
N-Boc-3-cyano-L-phenylalanine

Purity: 95% CAS No.: 131980-30-8 Molecular Formula: C15H18N2O4 Formula Weight: 290.31 Physical Form: Solid UN No.: UN3439 MDL No.: MFCD00797560

Formel: C₁₅H₁₈N₂O₄
Molecular Weight: 290,319 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00797560
CAS-Nummer: 131980-30-8
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR