2-Fluoro-3,4-methylenedioxybenzeneboronic acid pinacol ester 96%

Lieferant: Thermo Scientific

Synonyme: 2-Fluoro-3,4-methylenedioxyphenylboronic acid pinacol ester

H62587.03
H62587.03EA 577 EUR
H62587.03
2-Fluoro-3,4-methylenedioxybenzeneboronic acid pinacol ester 96%
2-Fluoro-3,4-methylenedioxybenzeneboronic acid pinacol ester

Purity: 96% Molecular Formula: C13H16BFO4 Formula Weight: 266.07 Physical Form: Solid

Formel: C₁₃H₁₆BFO₄
Molecular Weight: 266,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23098820
CAS-Nummer: 1429340-48-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR