3-Fluoro-5-nitrobenzeneboronic acid pinacol ester 96%

Lieferant: Thermo Fisher Scientific

H62652.03
H62652.03EA 334 EUR
H62652.03
3-Fluoro-5-nitrobenzeneboronic acid pinacol ester 96%
3-Fluoro-5-nitrobenzeneboronic acid pinacol ester

Purity: 96% CAS No.: 710348-94-0 Molecular Formula: C12H15BFNO4 Formula Weight: 267.06 Physical Form: Solid

Formel: C₁₂H₁₅BFNO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD22419276
CAS-Nummer: 710348-94-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR