4-Amino-3,5-difluorobenzeneboronic acid pinacol ester 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Amino-2,6-difluorophenylbronic acid, pinacol ester, 4-Amino-3,5-difluorophenylboronic acid pinacol ester

H62654.03
H62654.03EA 298 EUR
H62654.03
4-Amino-3,5-difluorobenzeneboronic acid pinacol ester 96%
4-Amino-3,5-difluorobenzeneboronic acid pinacol ester

Purity: 96% Molecular Formula: C12H16BF2NO2 Formula Weight: 255.07 Physical Form: Solid Appearance: White MDL No.: MFCD18434474

Formel: C₁₂H₁₆BF₂NO₂
Molecular Weight: 255,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18434474
CAS-Nummer: 939968-08-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR