3'-Brom-2'-fluoracetophenon 96%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-(3-Bromo-2-fluorophenyl)ethanone
H62708.03
H62708.03EA
209
EUR
H62708.03
3'-Brom-2'-fluoracetophenon 96%
3'-Brom-2'-fluoracetophenon
Purity: 96%
CAS No.: 161957-61-5
Molecular Formula: C8H6BrFO
Formula Weight: 217.04
Physical Form: Solid
Formel:
C₈H₆BrFO Molecular Weight: 217,04 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09264507 CAS-Nummer: 161957-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...