2-Fluor-4-nitrophenylboronsäurepinakolester 96%

Lieferant: Thermo Fisher Scientific

H62718.03
H62718.03EA 337 EUR
H62718.03
2-Fluor-4-nitrophenylboronsäurepinakolester 96%
2-Fluor-4-nitrophenylboronsäurepinakolester

Purity: 96% CAS No.: 1073353-89-5 Molecular Formula: C12H15BFNO4 Formula Weight: 267.06 Physical Form: Solid

Formel: C₁₂H₁₅BFNO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD09264075
CAS-Nummer: 1073353-89-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR