2-Cyan-4-fluorphenylboronsäurepinakolester 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H62729.MD
H62729.MDEA
63.5
EUR
H62729.MD
2-Cyan-4-fluorphenylboronsäurepinakolester 96%
2-Cyan-4-fluorphenylboronsäurepinakolester
Purity: 96%
CAS No.: 461451-63-8
Molecular Formula: C13H15BFNO2
Formula Weight: 247.07
Physical Form: Solid
Formel:
C₁₃H₁₅BFNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09260444 CAS-Nummer: 461451-63-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...