Z-Dab(Boc)-OH·DCHA 98%

Lieferant: Thermo Fisher Scientific

Synonyme: (S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt

H62772.03 H62772.06
H62772.03EA 91.8 EUR
H62772.03 H62772.06
Z-Dab(Boc)-OH·DCHA 98%
Z-Dab(Boc)-OH·DCHA

Purity: 98% CAS No.: 3350-13-8 Molecular Formula: C17H24N2O6.C12H23N Formula Weight: 553.70 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: Off-white MDL No.: MFCD00237341

Formel: C₂₉H₄₇N₃O₆
Molecular Weight: 533.71 g/mol
MDL: MFCD00237341
CAS-Nummer: 3350-13-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR