3-Fluor-4-(trifluormethyl)phenylboronsäurepinakolester 96%

Lieferant: Thermo Scientific

Synonyme: 2-(3-Fluoro-4-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Fluoro-4-(trifluoromethyl)benzeneboronic acid pinacol ester

H62811.03
H62811.03EA 369 EUR
H62811.03
3-Fluor-4-(trifluormethyl)phenylboronsäurepinakolester 96%
3-Fluor-4-(trifluormethyl)phenylboronsäurepinakolester

Purity: 96% CAS No.: 445303-67-3 Molecular Formula: C13H15BF4O2 Formula Weight: 290.06 Physical Form: Solid

Formel: C₁₃H₁₅BF₄O₂
Molecular Weight: 290,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878544
CAS-Nummer: 445303-67-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR