Boc-D-DAB(Z)-OH·DCHA 98%
Lieferant: Thermo Scientific
Synonyme:
Boc-D-Dab(Z)-OH·DCHA, (R)-4-Benzyloxycarbonylamino-2-(Boc-amino)butyric acid dicyclohexylammonium salt
H62817.06
H62817.06EA
322
EUR
H62817.06
Boc-D-DAB(Z)-OH·DCHA 98%
Boc-D-DAB(Z)-OH·DCHA
Purity: 98%
CAS No.: 101854-42-6
Molecular Formula: C17H24N2O6.C12H23N
Formula Weight: 533.70
Physical Form: Solid
MDL No.: MFCD00798628
Formel:
C₂₉H₄₇N₃O₆ Molecular Weight: 533,71 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00798628 CAS-Nummer: 101854-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...