2-Carbamoyl-3-fluorobenzeneboronic acid pinacol ester 96%
Lieferant: Thermo Fisher Scientific
H62904.MD
H62904.03
H62904.MDEA
157
EUR
H62904.MD
H62904.03
2-Carbamoyl-3-fluorobenzeneboronic acid pinacol ester 96%
2-Carbamoyl-3-fluorobenzeneboronic acid pinacol ester
Purity: 96%
Molecular Formula: C13H17BFNO3
Formula Weight: 265.09
Physical Form: Solid
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...